Toluene ≥98%, TECHNICAL
950 L
: Methylbenzene, Phenylmethane, FM approval
- :108-88-3
- :C₆H₅CH₃
- :MFCD00008512
- :92.14 g/mol
- :13,09607
- :110.6 °C (1013 hPa)
- :–95 °C
- :0.867 g/cm³ (20 °C)
- :4 °C
- :Ambient
- :203-625-9
- :1294
- :3,II
- :01-2119471310-51
Documents
30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC28701.364_SPEC_EN.pdf P20_VWRC28701.466_SPEC_EN.pdf P20_VWRC28701.550_SPEC_EN.pdf spec_VWRC28701.364 spec_VWRC28701.466
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
- Assay:Min. 98 %
- Pk:950 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.