IODOBROMIDE TS 1 * 100 mL
Catalog # CMPLUSP233
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test strips, peracetic acid, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach