ANTI-MAGE A10 CD8+ T CELLS, 1 TO 2 MILLION CELLS/VIAL, CRYOPRESERVED 1 * 2 Mou
ASTC-1125
ANTI-MAGE A10 CD8+ T CELLS, 1 TO 2 MILLION CELLS/VIAL, CRYOPRESERVED 1 * 2 Mou
ASTC-1125
Frequently Bought Together
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
: VWR Collection
Test tubes
: Greiner Bio-One
VWR®, Sampling Spoons
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Blender Bags
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Adhesive PCR Film Seals
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF