Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION 7.413 25C 1 * 500 mL
Catalog # ACSDPH018.L05
Supplier:
CPACHEM
Expand image
BUFFER SOLUTION 7.413 25C 1 * 500 mL
Catalog # ACSDPH018.L05
Supplier:
CPACHEM
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals