ContactUsLinkComponent
HighPurS-CERT - Potassium Nitrate (KNO3) - min. 99.999%, ISO 17034 and ISO/IEC 17025 certified CRM, ISO 17034 and ISO 17025 Conservation 36 months. (18 months after opening) Store under normal laboratory conditions, at temperatures between 15°C to 25 1 * 50 g
Catalog # CMPLPSKNC.50G
HighPurS-CERT - Potassium Nitrate (KNO3) - min. 99.999%, ISO 17034 and ISO/IEC 17025 certified CRM, ISO 17034 and ISO 17025 Conservation 36 months. (18 months after opening) Store under normal laboratory conditions, at temperatures between 15°C to 25 1 * 50 g
Catalog # CMPLPSKNC.50G
Frequently Bought Together