Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples. 1 * 10 Vial
Catalog # OXOISR0252E
Supplier:
THERMO FISHER DIAGNOSTICS
Expand image
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples. 1 * 10 Vial
Catalog # OXOISR0252E
Supplier:
THERMO FISHER DIAGNOSTICS
Frequently Bought Together
NBB®-A, Agar
Supplier: DOEHLER GROUP
VWR Chemicals, Base Materials for Dehydrated Media
Supplier: VWR Chemicals
SRK® Surfaces and Equipment Microbial Testing Devices
Supplier: Copan
VWR® Contact Plates, Blister Packed, Irradiated
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
Accessories for Anaerobic Systems, Oxoid
Supplier: THERMO FISHER DIAGNOSTICS