ContactUsLinkComponent
Trinocular tube 100% M-series - Beampath switchable 100% camera or 100% eyepieces - Interpupillary distance 51-77mm - Observation angle 30 - For High-end stereo microscopes - Low viewing height - Improved viewing conditions for photographing in com 1 * 1 items
Catalog # LEIM10450043
Supplier:  LEICA MICROSYSTEMS
Trinocular tube 100% M-series - Beampath switchable 100% camera or 100% eyepieces - Interpupillary distance 51-77mm - Observation angle 30 - For High-end stereo microscopes - Low viewing height - Improved viewing conditions for photographing in com 1 * 1 items
Catalog # LEIM10450043
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together