ContactUsLinkComponent
Trinocular tube 50% - Fixed beamsplitter 50% eyepieces and 50% camera - Interpupillary distance 51-77mm - Observation angle 30 - For High-end stereo microscopes - Low viewing height - Improved viewing conditions for photographing in combination wit 1 * 1 items
Catalog # LEIM10450042
Supplier:  LEICA MICROSYSTEMS
Trinocular tube 50% - Fixed beamsplitter 50% eyepieces and 50% camera - Interpupillary distance 51-77mm - Observation angle 30 - For High-end stereo microscopes - Low viewing height - Improved viewing conditions for photographing in combination wit 1 * 1 items
Catalog # LEIM10450042
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together