ContactUsLinkComponent
Sontara EC Green White 325x420mm multipurpose nonwoven wipe, 100% biodegradable and compostable nonwoven wipe, OK Compost Industrial and FSC® Mix certified and Seedling logo Strong, durable, absorbent, low-linting, 300 wipes per box. 1 * 300 items
Catalog # SONTD20000895
Supplier:  SONTARA
Sontara EC Green White 325x420mm multipurpose nonwoven wipe, 100% biodegradable and compostable nonwoven wipe, OK Compost Industrial and FSC® Mix certified and Seedling logo Strong, durable, absorbent, low-linting, 300 wipes per box. 1 * 300 items
Catalog # SONTD20000895
Supplier:  SONTARA
Frequently Bought Together