ContactUsLinkComponent
Sontara EC White, 305x420mm multipurpose nonwoven wipe: Strong, durable, absorbent, low-linting, 250 wipes per box. 1 * 250 items
Catalog # SONTD13791257
Supplier:  SONTARA
Sontara EC White, 305x420mm multipurpose nonwoven wipe: Strong, durable, absorbent, low-linting, 250 wipes per box. 1 * 250 items
Catalog # SONTD13791257
Supplier:  SONTARA
Frequently Bought Together