2-(1-METHYL-5-OXOPYRROLIDIN-3-YL)ACETICACID 1 * 50 g
Catalog # PHBLPBZ6042-50G
Supplier: Pharmablock (USA)
2-(1-METHYL-5-OXOPYRROLIDIN-3-YL)ACETICACID 1 * 50 g
Catalog # PHBLPBZ6042-50G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
1-Butanol ≥99.4% ACS
Supplier: Thermo Fisher Scientific