Water Cooling Kit for BIOne 1250 Controller. Requires One Kit Per Side 1 * 1 items
Catalog # DISI2800-3008
Supplier: DISTEK
Water Cooling Kit for BIOne 1250 Controller. Requires One Kit Per Side 1 * 1 items
Catalog # DISI2800-3008
Supplier: DISTEK
Frequently Bought Together
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
Titration beakers
Supplier: Mettler - Toledo
VWR®, Digital Calipers
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Conductivity standards
Supplier: WTW
Accessories for titrators
Supplier: Mettler - Toledo
Multi-parameter meters, handheld, 3320
Supplier: WTW
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
VWR®, Hand Tally Counter
Supplier: VWR Collection
Plastic cuvette for LICO
Supplier: Hach
Melting point and boiling point capillaries and tubes
Supplier: BUCHI