LITHIUM HEXAFLUOROPHOSPHATE SOLUTION 1 * 100 mL
SIAL901685-100ML
: Merck
Frequently Bought Together
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem