4,4,5,5-Tetramethyl-2-(4-(1,2,2-triphenylvinyl)phenyl)-1,3,2-dioxaborolane,98% mix TBC as stabilizer 1 * 0,25 g
Catalog # AMBDA1149541-250MG
Supplier: AMBEED
4,4,5,5-Tetramethyl-2-(4-(1,2,2-triphenylvinyl)phenyl)-1,3,2-dioxaborolane,98% mix TBC as stabilizer 1 * 0,25 g
Catalog # AMBDA1149541-250MG
Supplier: AMBEED
Frequently Bought Together
Dichloromethane stabilised for residue analysis, for pesticide residue analysis
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Tetrachloroethylene 99%, extra pure
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
1,2-Dichloroethane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific