Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Methyl 3,5-dibromobenzoate,98% 1 * 10 g
Catalog # AMBDA163152-10G
Supplier:
AMBEED
Expand image
Methyl 3,5-dibromobenzoate,98% 1 * 10 g
Catalog # AMBDA163152-10G
Supplier:
AMBEED
Frequently Bought Together
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific