1-Bromododecane,99% 1 * 500 g
Catalog # AMBDA293791-500G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Methylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Toluene ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific