Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate,98% 1 * 5 g
Catalog # AMBDA944088-5G
Supplier:
AMBEED
Expand image
Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate,98% 1 * 5 g
Catalog # AMBDA944088-5G
Supplier:
AMBEED
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99.5% for HPLC
Supplier: Thermo Fisher Scientific
1,4-Dioxane 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals