Acenaphthylen-1(2H)-one,98% 1 * 1 g
Catalog # AMBDA479417-1G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 100…120 °C, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, Biograde for DNA and peptides synthesis
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific