Dodec-11-yn-1-ol,98% 1 * 5 g
Catalog # AMBDA267431-5G
Supplier: AMBEED
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
n-Pentane, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Hexane (mixture of isomers) 98+%, extra pure
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
p-Xylene 99%, pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific