Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(R)-1,2,3,4-Tetrahydronaphthalen-1-amine,98% 1 * 25 g
Catalog # AMBDA236877-25G
Supplier:
AMBEED
Expand image
(R)-1,2,3,4-Tetrahydronaphthalen-1-amine,98% 1 * 25 g
Catalog # AMBDA236877-25G
Supplier:
AMBEED
Frequently Bought Together
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Toluene ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Ethyl Acetate, 99.5% for synthesis 1 * 5 L
Supplier: PanReac AppliChem