2-Iodo-1-methyl-4-nitrobenzene,98% 1 * 25 g
Catalog # AMBDA181575-25G
Supplier: AMBEED
Frequently Bought Together
Heptane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Diethylene glycol monobutyl ether ≥98%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
1-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone 99+%, extra pure
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane ≥99%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Supplier: Thermo Fisher Scientific