0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine,98% 1 * 1 g
AMBDA269028-1G
:
AMBEED
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine,98% 1 * 1 g
AMBDA269028-1G
:
AMBEED
Frequently Bought Together
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
: VWR Chemicals
Tetrahydrofuran ≥99.5% (by anhydrous basis) stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals