Thieno[3,2-b]thiophene-2-carboxylic acid,98% 1 * 0,1 g
Catalog # AMBDA264990-100MG
Supplier: AMBEED
Frequently Bought Together
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
Glycerol, 99% for synthesis 1 * 1.000 mL
Supplier: PanReac AppliChem
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Bromine ≥99.8%, liquid
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 35…60 °C ACS
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Methyl formate 98% for spectroscopy
Supplier: Thermo Fisher Scientific
p-Xylene 99%, pure
Supplier: Thermo Fisher Scientific