1,8-Dibromonaphthalene,98% 1 * 10 g
Catalog # AMBDA410357-10G
Supplier: AMBEED
Frequently Bought Together
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Supplier: VWR Chemicals
Chloroform
Supplier: Roth Carl
Chloroform 99+% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane stabilised for residue analysis, for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.7% for spectrophotometry
Supplier: Thermo Fisher Scientific
n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific