Bis[(pinacolato)boryl]methane,95% 1 * 25 g
AMBDA218916-25G
: AMBEED
Frequently Bought Together
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
: PanReac AppliChem
Glycerine 85% Ph. Eur.
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific
Water, BAKER ANALYZED®, J.T.Baker®
: Avantor