Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
N-Phenyldibenzo[b,d]furan-4-amine,99% 1 * 1 g
Catalog # AMBDA502928-1G
Supplier:
AMBEED
Expand image
N-Phenyldibenzo[b,d]furan-4-amine,99% 1 * 1 g
Catalog # AMBDA502928-1G
Supplier:
AMBEED
Frequently Bought Together
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
1-METHYL-2-PYRROLIDINONE 1 * 1 L
Supplier: Merck
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
KOLLIPHOR RH 40 1 * 1 kg
Supplier: Merck