(4-(Azetidin-1-ylsulfonyl)phenyl)boronic acid,98% 1 * 0,1 g
Catalog # AMBDA848136-100MG
Supplier: AMBEED
(4-(Azetidin-1-ylsulfonyl)phenyl)boronic acid,98% 1 * 0,1 g
Catalog # AMBDA848136-100MG
Supplier: AMBEED
Frequently Bought Together
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
ENZYMATIC YEAST BETA -GLUCAN KIT 1 * 50 Tests
Supplier: MEGAZYME
Methanol 99.9% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals