(4-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenyl)boronic acid,98% 1 * 0,25 g
AMBDA690231-250MG
: AMBEED
(4-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenyl)boronic acid,98% 1 * 0,25 g
AMBDA690231-250MG
: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals