Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-2-carboxylate,98% 1 * 0,25 g
AMBDA617405-250MG
:
AMBEED
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-2-carboxylate,98% 1 * 0,25 g
AMBDA617405-250MG
:
AMBEED
Frequently Bought Together
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
1,2-Dichlorobenzene, extra dry 98+%, AcroSeal®
: Thermo Fisher Scientific
Diethyl Ether stabilized with &sim 6 ppm of BHT (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 mL
: PanReac AppliChem
ASSAY VOLLSTÄNDIGE ZELLE ABLÖSUNG REAGEN
: DISCOVERX
Benzyl alcohol ≥97%, TECHNICAL
: VWR Chemicals
4,4'-Bis(dimethylamino)diphenylmethane 98%
: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
: Merck