Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(3-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid,95% 1 * 0,25 g
Catalog # AMBDA763595-250MG
Supplier:
AMBEED
Expand image
(3-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid,95% 1 * 0,25 g
Catalog # AMBDA763595-250MG
Supplier:
AMBEED
Frequently Bought Together
Chloroform
Supplier: Roth Carl
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water ACS
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chloroform stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals