(2-((3-Chlorobenzyl)oxy)phenyl)boronic acid,98% 1 * 0,1 g
Catalog # AMBDA661309-100MG
Supplier: AMBEED
(2-((3-Chlorobenzyl)oxy)phenyl)boronic acid,98% 1 * 0,1 g
Catalog # AMBDA661309-100MG
Supplier: AMBEED
Frequently Bought Together
1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Xylene (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether 99+% stabilised, pure
Supplier: Thermo Fisher Scientific