tert-Butyl 4-acetylpiperidine-1-carboxylate,98% 1 * 10 g
Catalog # AMBDA298596-10G
Supplier: AMBEED
Frequently Bought Together
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Iodomethane 99% stabilised
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
CYCLOPENTANE REAGENT GRADE 98% 1 * 500 mL
Supplier: Merck
Toluene ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Diethyl ether, anhydrous ≥99% stabilised ACS
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific