4-Bromo-2-chloro-1-(trifluoromethoxy)benzene,98% 1 * 25 g
Catalog # AMBDA413382-25G
Supplier: AMBEED
4-Bromo-2-chloro-1-(trifluoromethoxy)benzene,98% 1 * 25 g
Catalog # AMBDA413382-25G
Supplier: AMBEED
Frequently Bought Together
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Supplier: VWR Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Chloroform
Supplier: Roth Carl
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
2-Propanol solution in water 70%, technical
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific