Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
tert-Butyl (2-amino-2-iminoethyl)carbamate hydrochloride,95% 1 * 5 g
Catalog # AMBDA627256-5G
Supplier:
AMBEED
Expand image
tert-Butyl (2-amino-2-iminoethyl)carbamate hydrochloride,95% 1 * 5 g
Catalog # AMBDA627256-5G
Supplier:
AMBEED
Frequently Bought Together
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide 99.7%, pure
Supplier: Thermo Fisher Scientific
Chloroform
Supplier: Roth Carl
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 98.5+% ACS
Supplier: Thermo Fisher Scientific