0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
(4-(Difluoromethoxy)phenyl)boronic acid,95% 1 * 0,25 g
AMBDA212028-250MG
:
AMBEED
(4-(Difluoromethoxy)phenyl)boronic acid,95% 1 * 0,25 g
AMBDA212028-250MG
:
AMBEED
Frequently Bought Together
Dichloromethane stabilised Ph. Eur.
: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Water, pure, demineralised
: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
Methanol bio-based ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Chlorobenzene 99.9% for HPLC
: Thermo Fisher Scientific
Methyl ethyl ketone, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
Butyl acetate ACS
: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
: Thermo Fisher Scientific