Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Thianthren-1-ylboronic acid,97% 1 * 0,25 g
Catalog # AMBDA184733-250MG
Supplier:
AMBEED
Expand image
Thianthren-1-ylboronic acid,97% 1 * 0,25 g
Catalog # AMBDA184733-250MG
Supplier:
AMBEED
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
ISOPENTYL ACETATE 98% 1 * 100 mL
Supplier: Merck
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific