tert-Butyl (6-aminohexyl)carbamate hydrochloride,98% 1 * 5 g
AMBDA360427-5G
: AMBEED
Frequently Bought Together
Ethyl acetate ≥99%, TECHNICAL
: VWR Chemicals
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
: VWR Chemicals