Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
2-(Adamantan-1-yl)acetic acid,98% 1 * 1 g
Catalog # AMBDA139949-1G
Supplier:
AMBEED
Expand image
2-(Adamantan-1-yl)acetic acid,98% 1 * 1 g
Catalog # AMBDA139949-1G
Supplier:
AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
Diethyl ether stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Supplier: Avantor
Ethanolamine 99%
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor