(S)-1-(Naphthalen-2-yl)ethanol,98% 1 * 0,1 g
AMBDA325397-100MG
: AMBEED
Frequently Bought Together
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
Ethylene glycol ≥99%
: Thermo Fisher Scientific
n-Hexane ≥95% for spectrophotometry
: Thermo Fisher Scientific
Glycerine, Hi-LR™
: HIMEDIA
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Isobutanol 99+% for HPLC
: Thermo Fisher Scientific