3-Butyl-1-vinyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide,98% mix TBC as stabilizer 1 * 25 g
Catalog # AMBDA1174281-25G
Supplier: AMBEED
3-Butyl-1-vinyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide,98% mix TBC as stabilizer 1 * 25 g
Catalog # AMBDA1174281-25G
Supplier: AMBEED
Frequently Bought Together
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Paraffin, liquid, liquid Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Acetone, BAKER, J.T.Baker®
Supplier: Avantor