tert-Butyl (2-vinylcyclopropyl)carbamate,97% mix TBC as stabilizer 1 * 0,1 g
Catalog # AMBDA1153649-100MG
Supplier: AMBEED
tert-Butyl (2-vinylcyclopropyl)carbamate,97% mix TBC as stabilizer 1 * 0,1 g
Catalog # AMBDA1153649-100MG
Supplier: AMBEED
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals