0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
4-(Trifluoromethyl)pyridine 1-oxide,95% 1 * 0,25 g
AMBDA636698-250MG
:
AMBEED
4-(Trifluoromethyl)pyridine 1-oxide,95% 1 * 0,25 g
AMBDA636698-250MG
:
AMBEED
Frequently Bought Together
Chloroform, extra dry 99.9% stabilised, AcroSeal®
: Thermo Fisher Scientific
Toluene ≥98%, TECHNICAL
: VWR Chemicals
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific
4-Methyl-2-pentanone 98.5+% ACS
: Thermo Fisher Scientific
CYCLOPENTANE REAGENT GRADE 98% 1 * 500 mL
: Merck