Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pycnometer acc. Gay-Lussac, precisely adjusted to the third place of decimal, acc.to DIN 12 797, 2 ml, with certificate 1 * 1 items
Catalog # WITG3902002IC
Supplier:
witeg Labortechnik
Expand image
Pycnometer acc. Gay-Lussac, precisely adjusted to the third place of decimal, acc.to DIN 12 797, 2 ml, with certificate 1 * 1 items
Catalog # WITG3902002IC
Supplier:
witeg Labortechnik
Frequently Bought Together
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Dropping funnels, cylindrical, with pressure balance
Supplier: Lenz Laborglas GmbH & CO.KG
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Beakers, low form, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Low Form
Supplier: VWR Collection