Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TRYPTONE YEAST EXTRACT BROTH 1 * 500 g
Catalog # HIMEM356-500G
Supplier:
HIMEDIA
Expand image
TRYPTONE YEAST EXTRACT BROTH 1 * 500 g
Catalog # HIMEM356-500G
Supplier:
HIMEDIA
Frequently Bought Together
VWR® Contact Plates, Blister Packed, Irradiated
Supplier: VWR Chemicals
SRK® Surfaces and Equipment Microbial Testing Devices
Supplier: Copan
Aerobic count plates, Petrifilm™
Supplier: 3M Food Safety
NBB®-A, Agar
Supplier: DOEHLER GROUP
VWR®, Fluids and culture media for sterility testing of pharmaceutical products
Supplier: VWR Chemicals
VWR®, Contact slides
Supplier: VWR Chemicals
VWR® Catalase Reagent
Supplier: VWR Chemicals
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals