ContactUsLinkComponent
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 100, 2.7 um, PEEK-lined. Specifications:- Endcapped: No. Guard Column: No. Hardware: PEEK L ined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C 1 * 1 items
Catalog # HEWL675775-924
Supplier:  Agilent
undefined
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 100, 2.7 um, PEEK-lined. Specifications:- Endcapped: No. Guard Column: No. Hardware: PEEK L ined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C 1 * 1 items
Catalog # HEWL675775-924
Supplier:  Agilent
Frequently Bought Together