Orders LinkContactUsLinkComponent
Static dissipative facemasks
Static dissipative facemasks
L/XL, White
Catalog # DESC97554
Supplier:   DESCO
Static dissipative facemasks
Catalog # DESC97554
Supplier:  DESCO
Static dissipative facemasks
L/XL, White
Price
23,40 €
Log in for availability
1 * 1 items
Product Details & Documents
These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofiliment.
  • Can be washed multiple times in a mild detergent
  • Elastic ear loops
  • Adjustable nose wire helps ensure mask fits securely
  • Acceptable for use in ESD protected areas
  • Provides continuous and consistent charge dissipation
Elastic bands range from small-medium (69,85 mm) to large-extra large (82,55 mm). The mask area is the same size for all items.
Specifications
  • Colour:
    White
  • Seq:
    05
  • Size:
    L/XL