Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
Green
Catalog # KART0291912
Supplier:   KARTELL
Embedding rings
Catalog # KART0291912
Supplier:  KARTELL
Embedding rings
Embedding rings
Green
Price
317,00 €
Log in for availability
1 * 2.000 items
Product Details & Documents
Embedding rings made from ABS.
  • Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.
Specifications
  • Colour:
    Green