Orders LinkContactUsLinkComponent
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
Micro test tube, graduated, 1,5 ml, Green, PP
Catalog # KART0027912
Supplier:   KARTELL
Micro Test Tubes, Graduated
Catalog # KART0027912
Supplier:  KARTELL
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
Micro test tube, graduated, 1,5 ml, Green, PP
Price
299,00 €
Log in for availability
1 * 10.000 items
Product Details & Documents
Ideal for samples analysis.
  • Made from PP
  • Flat cap with writing area
  • Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Certifications: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.
Specifications
  • Capacity:
    1,5 ml
  • Colour:
    Green
  • Description:
    Micro test tube, graduated
  • Material:
    PP
  • Seq:
    03