Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Contact slide Chrom 3, 20
Catalog # LIOF525462
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF525462
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Contact slide Chrom 3, 20
CAS Number:  
Price
62,50 €
Log in for availability
1 * 20 items
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pk:
    20
  • Description:
    Contact slide Chrom 3
Specifications