PIPET TIPS FLEXSTATION 96-WELL CLEAR 1 * 10 items
Catalog # MLDV9000-0912
Supplier: Molecular Devices
PIPET TIPS FLEXSTATION 96-WELL CLEAR 1 * 10 items
Catalog # MLDV9000-0912
Supplier: Molecular Devices
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Accessories for Mettler balances
Supplier: Mettler - Toledo
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Cell culture media, DMEM-Ham's F12
Supplier: Biowest
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
Hygromycin B in solution
Supplier: PanReac AppliChem